missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Spinosad, mixture of A and D

Product Code. p-7214730 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17251316

Brand: Thermo Scientific Chemicals 467190010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 168316-95-8 | C42H71NO9 | 734.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168316-95-8

Specifications

Quantity 1 g
Melting Point 142.0°C to 145.0°C
Molecular Formula C42H71NO9
InChI Key RQOIAWYOVOXMST-UHFFFAOYNA-N
SMILES CCC1CCCC(OC2CCC(C(C)O2)N(C)C)C(C)C(=O)C2CC3C4CC(CC4CCC3C2CC(=O)O1)OC1CC(C)C(OC)C(OC)C1OC
IUPAC Name 13-{[5-(dimethylamino)-6-methyloxan-2-yl]oxy}-9-ethyl-14-methyl-2-[(2,3,4-trimethoxy-5-methylcyclohexyl)oxy]-icosahydro-1H-as-indaceno[3,2-d]oxacyclododecane-7,15-dione
Molecular Weight (g/mol) 734.03
Chemical Name or Material Spinosad
compliance-icons
Product Identifier
  • Spinosad
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Spinosad

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.