missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPINLW1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596071
This item is not returnable.
View return policy
Description
Immunogen sequence: PGLTDWLFPR RCPKIREECE FQERDVCTKD RQCQDNKKCC VFSCGKKCLD LKQDVCEMPK ETGPCLAYFL HWWYDKKDNT CSMFVYGGCQ GNNNNFQSKA NCLNTCKNKR FP; Positive Samples: Mouse testis; Cellular Location: Cell surface.
Eppin, an epididymal protease inhibitor, belongs to the WFDC family and to the telomeric cluster. The gene encoding the protein is localized to chromosome 20q12-q13 in centromeric and telomeric clusters. It expresses three mRNAs encoding two isoforms of a cystine-rich protein that contains Kunitz-type and WAP-type (four disulfide core) protease inhibitor consensus sequences. The mouse gene lies in a cluster of putative Eppin-like genes on mouse chromosome 2. Following ejaculation, Eppin is bound to semenogelin in seminal plasma and on human spermatozoa. This complex of Eppin and Semenogelin can provide antimicrobial activity for spermatozoa. It can also provide for the preparation and survival of spermatozoa for fertility in the female reproductive tract. Eppin, which is a secreted protein, is expressed in epididymis and testis.
Specifications
| SPINLW1 | |
| Polyclonal | |
| Unconjugated | |
| EPPIN | |
| 1700024E17Rik; cancer/testis antigen 71; cancer/testis antigen 72; CT71; CT72; dJ461P17.2; epididymal peptidase inhibitor; epididymal protease inhibitor; EPPIN; EPPIN1; EPPIN2; EPPIN3; protease inhibitor WAP7; serine peptidase inhibitor-like, with Kunitz and WAP domains 1 (eppin); serine protease inhibitor-like with Kunitz and WAP domains 1; serine protease inhibitor-like, with Kunitz and WAP domains 1 (eppin); Spinlw1; WAP four-disulfide core domain protein 7; WAP four-disulfide core domain protein 8; WAP7; WFDC7; Wfdc8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57119, 685161, 75526 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.93 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| D4A2Z2, O95925, Q9DA01 | |
| EPPIN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-133 of human EPPIN (NP_0651311). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction