missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINK9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24661-1-AP-20UL
This item is not returnable.
View return policy
Description
SPINK9 is a serine protease inhibitor which specifically inhibits KLK5. It may contribute to the regulation of the desquamation process in skin by inhibiting KLK5.Specifications
| SPINK9 | |
| Polyclonal | |
| Unconjugated | |
| SPINK9 | |
| LEKTI2, SPINK9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 643394 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5DT21 | |
| SPINK9 | |
| SPINK9 Fusion Protein Ag19924 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction