missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPINK7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551287
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SPINK7 protein.
SPINK7 (serine peptidase inhibitor, Kazal type 7), also known as Ecg2 (esophagus cancer-related gene 2 protein), is a 76 amino acid secreted protein. Containing one Kazal-like domain, SPINK7 is thought to be a serine protease inhibitor. The gene that encodes SPINK7 maps to human chromosome 5, which contains 181 million base pairs encoding around 1,000 genes. Chromosome 5 is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome. Deletion of 5q or chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.Probable serine protease inhibitor.
Specifications
| SPINK7 | |
| Polyclonal | |
| Unconjugated | |
| SPINK7 | |
| ECG2; ECRG2; ECRG-2; EG408198; esophagus cancer related gene 2; esophagus cancer-related gene 2 protein; esophagus cancer-related gene-2; esophagus cancer-related protein 2; serine peptidase inhibitor, Kazal type 7; serine peptidase inhibitor, Kazal type 7 (putative); serine protease inhibitor Kazal-type 7; SPINK7; UNQ745/PRO1474 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84651 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P58062 | |
| SPINK7 | |
| Synthetic peptide of human SPINK7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction