missing translation for 'onlineSavingsMsg'
Learn More

SPINK1 Antibody Pair, Novus Biologicals™

Product Code. 18361759 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361759 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361759 Supplier Novus Biologicals Supplier No. H00006690AP41

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pair

SPINK1 Monoclonal antibody specifically detects SPINK1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPINK1
Applications Sandwich ELISA
Classification Monoclonal
Conjugate Unconjugated
Formulation 1x PBS, pH 7.4
Gene Alias pancreatic secretory trypsin inhibitor, PCTTSpink3, PSTISerine protease inhibitor Kazal-type 1, serine peptidase inhibitor, Kazal type 1, serine protease inhibitor, Kazal type 1, TATITumor-associated trypsin inhibitor
Quantity 1 Set
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6690
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.