missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINK1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98602AF594
This item is not returnable.
View return policy
Description
SPINK1 Polyclonal antibody specifically detects SPINK1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SPINK1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| pancreatic secretory trypsin inhibitor, PCTTSpink3, PSTISerine protease inhibitor Kazal-type 1, serine peptidase inhibitor, Kazal type 1, serine protease inhibitor, Kazal type 1, TATITumor-associated trypsin inhibitor | |
| Produced in rabbits immunized with E. coli-derived Human SPINK1 fragment. | |
| 0.1 mL | |
| Cancer | |
| 6690 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction