missing translation for 'onlineSavingsMsg'
Learn More

Spinetoram J, Dr. Ehrenstorfer

Product Code. 30309364
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309364 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309364 Supplier Dr. Ehrenstorfer Supplier No. DREC16972780
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Spinetoram J
CAS 187166-40-1
Analyte or Component Names Spinetoram J
Shipping Condition +20°C
Molecular Formula C42 H69 N O10
Certifications/Compliance ISO/IEC 17025
Synonym Spinetoram, DE 175J, Spinetoram J
InChI Formula InChI=1S/C42H69NO10/c1-10-27-13-12-14-35(53-37-18-17-34(43(6)7)24(4)49-37)23(3)38(45)33-21-31-29(32(33)22-36(44)51-27)16-15-26-19-28(20-30(26)31)52-42-41(47-9)40(48-11-2)39(46-8)25(5)50-42/h21,23-32,34-35,37,39-42H,10-20,22H2,1-9H3/t23-,24-,25+,26-,27+,28-,29-,30-,31-,32+,34+,35+,37+,39+,40-,41-,42+/m1/s1
SMILES CCO[C@@H]1[C@@H](OC)[C@H](C)O[C@@H](O[C@@H]2C[C@H]3CC[C@H]4[C@@H]5CC(=O)O[C@@H](CC)CCC[C@H](O[C@H]6CC[C@@H]([C@@H](C)O6)N(C)C)[C@@H](C)C(=O)C5=C[C@H]4[C@@H]3C2)[C@@H]1OC
Molecular Weight (g/mol) 748
Quantity 10 mg
Formula Weight 747.4921 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.