missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spinesin Antibody (OTI4A11), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74333PE
This item is not returnable.
View return policy
Description
Spinesin Monoclonal specifically detects Spinesin in Human samples. It is validated for Flow Cytometry.
Specifications
| Spinesin | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| TMPRSS5 | |
| Full length recombinant protein of human Spinesin (NP_110397) produced in HEK293T cells. | |
| 0.1 mL | |
| 80975.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI4A11 | |
| Flow Cytometry | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, MGC141886, MGC148044, Spinesin, transmembrane protease serine 5, transmembrane protease, serine 5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction