missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SPINeasy™ Virus RNA Kit

Product Code. 17884126 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 Preps
Unit Size:
50 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17884126 50 Preps 50 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17884126 Supplier MP Biomedicals™ Supplier No. 116537050
  • ONLINE EXCLUSIVE PRICE
    £260.00
    £243.60 / Pack of 50
    Save £16.40
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spin column technology for quick and easy isolation of viral RNA from cell culture media and bodily fluids in only 20 minutes. Highly purified RNA suitable for RT-PCR, qPCR, RNA-seq, and Northern Blot Analysis.

  • SPINeasy Virus RNA Kit is a silica membrane spin column kit that enables quick and convenient extraction of viral RNA from cell culture media and bodily fluids, such as saliva and serum, without the need for phenol or chloroform extractions.
  • Through a simple workflow, viral RNA is typically extracted within 20 minutes and is immediately available for downstream applications. including RT-PCR, qPCR, and RNA-Seq.

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Format Spin Column
Isolation Technology Spin Column
Sample Type Cell Culture Media and Bodily Fluids (Serum and Saliva); 100 to 500 µL of sample
Includes 30 mL Lysis Buffer VR, 6 mL Wash Buffer VR, 10 mL Nuclease-free water, 1 vial Carrier RNA, 50 x Column VR with collection tubes
Quantity 50 Preps
compliance-icons
Product Identifier
  • SPINeasy Virus RNA Kit, 50 preps
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Guanidine Hydrochloride

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.