missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals SPINeasy RNA Kit for Yeast

Product Code. 30281094 Shop All MP Biomedicals Products
Change view
Click to view available options
No. of Reactions:
50 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
30281094 50 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30281094 Supplier MP Biomedicals Supplier No. 116565050
  • ONLINE EXCLUSIVE PRICE
    £474.00
    £444.15 / Pack of 1
    Save £29.85
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SPINeasy RNA Kit for Yeast is a high-performance RNA extraction kit which is developed based on silica-membrane spin-column technology.

SPINeasy RNA Kit for Yeast is a high-performance RNA extraction kit which is developed based on silica-membrane spin-column technology. This kit employs a quick desalting technique to eliminate contaminating macromolecules while avoiding harmful chemical solvents, and spheroplasting to successfully release RNA through carbohydrate-containing cell walls, without the need of any enzymes. Up to 2 x 108 cells can be handled at once without any DNase treatment. The resulting RNA is of exceptional purity, free of proteins or DNA in eluted samples.

Usage Statement:

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Northern blotting
  • RT-PCR
  • Next-Generation Sequencing
  • Microarray
  • hybridization

Specifications

Format Kit
Isolation Technology Silica Spin Column
Sample Type Yeast
No. of Reactions 50 Preps
Product Type RNA Isolation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.