missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Produces high-integrity RNA without phenol or chloroform, immediately ready for real-time PCR, RT-PCR, and sequencing applications.
- RNA MATRIX is a proprietary silica-based binding matrix offering flexibility in RNA binding capacities and selectively binds RNA for purification.
- The RNA is preferentially purified from other cellular components such as proteins, without the use of phenol or chloroform.
- The resulting RNA is of high integrity and suitable for various downstream applications, including real-time PCR, reverse transcription PCR (RT-PCR), and sequencing.
Specifications
Specifications
| Buffer | Lysis Buffer |
| Control Type | RNA Control |
| Isolation Technology | Spin Column |
| Species | All |
| Sample Type | Urine |
| No. of Reactions | 50 Preps |
| Product Type | RNA Kit for Urine |
| Sterility | Non-sterile |
| Target | Total RNA |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?