Learn More
MP Biomedicals SPINeasy RNA Kit for Blood
Shop All MP Biomedicals ProductsDescription
The SPINeasy RNA Kit for Blood is an advanced RNA extraction solution leveraging state-of-the-art silica-membrane spin-column technology. Specifically designed for the isolation of RNA from whole blood preserved in various anticoagulants, this kit streamlines the process to under 15 minutes. RNA preservation is paramount, and the Lysis Buffer SL1 eliminates the need for excessive homogenization while ensuring immediate RNase inactivation, preventing RNA degradation.
Notably, the SPINeasy RNA Kit for Blood eliminates the requirement for prior erythrocyte lysis and leukocyte centrifugation, simplifying the RNA purification process.
The inclusion of Fast DNase facilitates efficient on-column DNA digestion, swiftly removing co-purified DNA bound to the silica membrane. Dual washing steps have been optimized to effectively eliminate:
- salts
- metabolites
- and large cellular components
contributing to a hassle-free and highly time-efficient RNA purification.
Following these steps, the elution of high-yield and pure RNA is achieved using RNase-free water.
The resulting RNA is well-suited for various downstream applications, including:
- PCR
- RT-qPCR amplification
- and sequencing.
Specifications
Specifications
| Isolation Technology | Silica Spin Column |
| Sample Type | Blood |
| No. of Reactions | 50 Preps |
| Product Type | RNA Isolation |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.