missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- The SPINeasy Host Depletion Microbial DNA Kit provides an easy-to-use workflow to isolate and enrich microbial DNA from samples containing high amounts of host DNA. Host DNA contamination is common in samples such as swabs and bodily fluids. The presence of high amounts of human DNA can limit the application and sensitivity of downstream shotgun metagenomic sequencing or microbiome analysis, as most of the extracted DNA is from human and not microbial origin.
- The SPINeasy Host Depletion Microbial DNA Kit provides efficient reduction of host DNA from these samples by selective lysis of host cells with our specially formulated Host Lysis Buffer. Host DNA, which has been released into the solution, is then removed from the supernatant through centrifugation. A further host depletion treatment provides enzymatic degradation of any remaining host DNA, as well as DNA coming from dead microbial cells. Finally, the remaining intact microbial cells are lysed through a combination of specially formulated buffers and highly efficient homogenization using FastPrep instruments from MP Biomedicals. Enriched microbial DNA is then purified using a convenient silica membrane spin-column technology workflow and ready for downstream molecular applications, such as PCR and NGS.
Specifications
Specifications
| Content And Storage | Store at Room Temperature, Dry Environment |
| Isolation Technology | Spin Column |
| Sample Type | Saliva, Swabs, Bodily Fluid |
| Includes | 12 mL Equilibration buffer,50x Lysing Matrix E, 60 mL Host Lysis Buffer, 6 mL Host Depletion Buffer, 1 Vial Host Depletion Enzyme HDE, 200 μL HDE Reconstitution Buffer, 12 mL Microbial Selection Buffer, 40 mL Microbial Lysis Buffer, 30 mL Wash Buffer HD1, 6 mL Wash Buffer HD2, 6 mL Wash Buffer HD, 50x Column HD with collection tubes |
| Quantity | 50 Preps |
Safety and Handling
Product Identifier
- SPINeasy Host Depletion Microbial DNA Kit
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Guanidine Hydrochloride, Manganese Sulfate Monohydrate, Calcium Chloride, TRITONX-100
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?