missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ SPINeasy™ DNA/RNA Kit for Soil
Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 Preps
Unit Size:
Each
Description
- The SPINeasy DNA/RNA Kit for Soil integrates our proprietary inhibitor removal and spin column technology for selective binding of DNA (blue column) and RNA (white column) from a single soil sample.
- Humic acids, heavy metals, and other aromatic components are among the most notorious PCR inhibitors found in soil samples, leading to false negative or inaccurate results.
- Non‐optimized nucleic acid extraction protocols often co‐purify inhibitors.
- The SPINeasy DNA/RNA Kit for Soil allows for fast and simultaneous isolation of pure microbial genomic DNA and RNA from challenging soil samples, including those with low biomass or those that are highly contaminated.
- The extracted nucleic acids lack contaminants and are suitable for use in downstream applications, such as qPCR, RT-qPCR, and sequencing, without the need for further inhibitor removal steps.
Specifications
Specifications
| Content And Storage | Store at Room Temperature, Dry Environment |
| Isolation Technology | Spin Column |
| Includes | Equilibration buffer (24 mL), Lysing Matrix YB (50 ea), Buffer SDR1A (18 mL), Buffer SDR1B (36 mL), Buffer SDR2 (24 mL), Buffer SDR3 (45 mL), Buffer SDR4 (45 mL), Buffer SDR5 (90 mL), Buffer SDR6 (150 mL), RNase Free Water (15 mL), DNA Column (50 ea), RNA Column (50 ea), 2 mL Microcentrifuge Tubes (100 ea), DNA Collection Tubes (250 ea), RNA Collection Tubes (150 ea), DNA Elution Tubes (50 ea), RNA Elution Tubes (50 ea) |
| Quantity | 50 Preps |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction