missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SPINeasy™ DNA Kit for Water

Product Code. 30281215 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 Preps
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30281215

Brand: MP Biomedicals™ 116536050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifically designed for the isolation of microbial DNA from various water samples. Fast and easy extraction of gDNA in less than 30 minutes. Supplied with filter membranes and Lysing matrix tubes to capture and concentrate microbes and efficiently homogenize in using a FastPrep instrument.

  • SPINeasy DNA Kit for Water is a high-performance water genomic DNA extraction kit based on silica membrane spin column technology.
  • The kit has been specifically designed to concentrate microbes present in water samples by filtration and transfer to a Lysing Matrix tube (4.5 mL) for the best extraction of microbial gDNA from water in less than 30 min.
  • Water samples are processed using our uniquely formulated Lysis Buffer W1 and Lysing Matrix E to effectively lyse various types of microbial cells, including bacteria, fungi, virus, and protozoans.
  • Column W1 (provided with the kit) has high binding capacity and selectivity for gDNA.
  • The combination of components in the kit helps extract microbial gDNA of high yield and purity that is ready for downstream applications, such as restriction digestion, PCR, and NGS.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Isolation Technology Spin Column
Sample Type Water Samples
Includes 50 x 5 mL Lysing Matrix E, 60 mL Lysis Buffer W1, 8 mL Lysis Buffer W2, 550 µL RNase A Solution, 15 mL Inhibitor Removal W, 30 mL Binding Buffer W, 9 mL Wash Buffer W1, 6 mL Wash Buffer W2, 10 mL DES Buffer, 50 x Filter Membrane, 50 x Column W1, 50 x 2.0 mL Collection Tubes, 50 x 1.5 mL Collection Tubes
Quantity 50 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.