missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP BIOMEDICALS SPINeasy DNA Kit for Saliva 50 Preps
Shop All MP Biomedicals Products
Click to view available options
:
50 Preps
Description
- The use of saliva as a source of DNA over blood samples has become an attractive approach for various applications ranging from genetic studies to pathogen detection.
- Unlike blood sampling, saliva collection is an:
- Easy
- Painless
- Non-invasive
- Method which does not require trained personnel for the collection process. In addition, saliva does not coagulate which makes it easier for handling than blood.
- However, storing saliva samples can be problematic.
- The SPINeasy DNA Kit for Saliva from MP Biomedicals makes saliva sampling easier by using our specially formulated Saliva Preservation Solution (SPS) to preserve the sample at room temperature without compromising the quality.
- SPINeasy DNA Kit for Saliva allows isolation of DNA from:
- Fresh
- Frozen
- SPS-preserved saliva that is extracted with a quick and easy protocol, using silica spin-column technology. Purified DNA is recovered in high yield and purity, suitable for various downstream molecular applications.
- Performance: SPINeasy DNA Kit for Saliva offers superior performance. The kit has been optimized and demonstrated competitive performance over competitor kits in both purity and yield.
- Usage Statement: Research Use Only (RUO).
- Isolate gDNA from 25 - 500 μL of saliva with simple procedures
- DNA ready to use for downstream application
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Silica Spin Column |
| Includes | 50 Preps |
| Product Type | DNA Extraction |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction