missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SPINeasy™ DNA Kit for Microbiome

Product Code. 30057357 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 Preps
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30057357

Brand: MP Biomedicals™ 116553050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For simple and fast isolation of microbial genomic DNA from various samples. Lysing Matrix E and specially formulated buffers for thorough and complete microbial lysis with FastPrep instruments.

  • SPINeasy DNA Kit for Microbiome allows for efficient and unbiased lysis of microbes, including gram positive and negative bacteria, fungi, protozoans, and viruses found in various swab, body fluid, stool, soil, and water samples.
  • This kit features a highly effective lysis system and silica membrane spin column technology. With the use of specially formulated Buffer MB1 and Lysing Matrix E in combination with FastPrep instruments from MP Biomedicals, efficient lysis of various samples can be achieved within seconds.
  • Provided with the kit, Column MB and kit buffers are designed to deliver gDNA of high yield and purity in fewer steps, ready for downstream applications such as qPCR, restriction digestion, and sequencing.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Isolation Technology Spin Column
Sample Type Swabs, Sputum, Body Fluids, Stool, and Environmental Samples (e.g. soil and water)
Includes Lysing Matrix E (50 ea), RNase A Solution (2 mL), Buffer MB1 (45 mL), Buffer MB2 (17 mL), Buffer MB3 (21 mL × 2), Buffer MB4 (28 mL), Buffer MB5 (20 mL × 2), Buffer MB6 (10 mL), Column MB (50 ea), 1.5 mL Collection Tubes (50 ea)
Quantity 50 Preps
compliance-icons compliance-icons
Product Identifier
  • SPINeasy DNA Kit for Microbiome
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Isopropanol, Guanidine Hydrochloride

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.