missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPIN4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560749
This item is not returnable.
View return policy
Description
Immunogen sequence: MSPPTVPPMG VDGVSAYLMK KRHTHRKQRR KPTFLTRRNI VG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 43%.
Exhibits H3K4me3-binding activity. [UniProt].
Specifications
| SPIN4 | |
| Polyclonal | |
| Unconjugated | |
| SPIN4 | |
| 9630042H07Rik; FLJ44984; MGC133224; novel Spin/Ssty repeat containing protein; Spin4; spindlin family member 4; spindlin family, member 4; spindlin-4; TDRD28 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 139886 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q56A73 | |
| SPIN4 | |
| Recombinant protein corresponding to Human SPIN4. Recombinant protein control fragment (Product #RP-96589). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction