missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPIN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576218
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Human SPIN2 expressed by 2 IL3-dependent murine myeloid cell lines was moderately antiapoptotic following IL3 removal, but it was unable to block apoptosis induced by the chemotherapy agent doxorubicin. On the contrary, SPIN2-expressing cells were more sensitive to doxorubicin-mediated cell death. Also, SPIN2 did not combat the proapoptotic effects of Fas ligand stimulation in the Jurkat human T-cell line. Deletion of the C-terminal amino acids of SPIN2 diminished its antiapoptotic activity. In the murine myeloid cells, SPIN2 overexpression increased the cell number doubling times and slowed their rate of growth. There was also an increased percentage of cells in G2/M, which was more pronounced following IL3 withdrawal.
Specifications
| SPIN2 | |
| Polyclonal | |
| Unconjugated | |
| SPIN2B | |
| BC068162; dJ323P24.2; Spin2; SPIN-2; SPIN2_duplicate; SPIN2B; SPIN-2B; Spin2c; SPIN-2C; Spindlin 2B; spindlin family member 2B; spindlin family, member 2; spindlin family, member 2 duplicate; spindlin family, member 2B; spindlin family, member 2C; spindlin-2; spindlin-2B; Spindlin-2C; spindlin-like protein 2; spindlin-like protein 2B; Spindlin-like protein 2C; TDRD26; telomeric SPIN2 copy | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 278240, 474343 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NVE3, Q9BPZ2 | |
| SPIN2B, Spin2c | |
| Recombinant protein of human SPIN2B. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion