missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPIN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584790
This item is not returnable.
View return policy
Description
Immunogen sequence: MKTPFGKTPG QRSRADAGHA GVSANMMKKR TSHKKHRSSV GPSKPVSQPR RNI Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.
SPIN1 (spindlin 1), also known as SPIN or ovarian cancer-related protein (OCR), is a 262 amino acid nuclear protein suggested to play a role in regulation of the cell-cycle during the transition from gamete to embryo. A member of the SPIN/STSY family, SPIN1 localizes to interphase nucleus and mitotic chromosomes, and is modified by phosphorylation in a cell-cycle-dependent fashion. A meiotic spindle-binding protein, SPIN1 overexpression has been shown to cause defects in mitotic spindles, thereby resulting in chromosome instability and potential tumorigenesis.
Specifications
| SPIN1 | |
| Polyclonal | |
| Unconjugated | |
| SPIN1 | |
| 30000 Mr metaphase complex; OCR; Ovarian cancer-related protein; SPIN; Spin1; spindlin 1; Spindlin1; spindlin-1; SSEC P; TDRD24 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10927 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y657 | |
| SPIN1 | |
| Recombinant protein corresponding to Human SPIN1. Recombinant protein control fragment (Product #RP-107652). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction