Learn More
Labconco™ SpillStopper™ Work Surface for 8ft. Protector™Premier™and Protector™XL Hoods
Product Code. 18724204 Shop All Labconco Corporation Products
Change view
Click to view available options
Cut Style:
Left and Right Rear Cupsink Cutouts
Unit Size:
Each
Product Code. Cut Style Unit Size
18724204 Left and Right Rear Cupsink Cutouts Each
1 options
Product Code. 18724204 Supplier Labconco™ Supplier No. 9500811
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
SpillStopper work surfaces are dished to contain spills
  • Molded from special formulation of corrosion-resistant epoxy resins
  • Front edge has large radius to aerodynamically direct airflow into the fume hood
  • Pre-drilled holes for venting and plumbing passthroughs are provided
  • Some include 6 x 3 in. cupsink cutouts
9500820 and 9500802
Not for use with Protector Solvent Storage Cabinets
Specifications
For Use With (Equipment) Protector XL Laboratory Hood
Product Type SpillStopper Work Surface (8 ft.)
For Use With (Application) 8 Ft. Protector Premier and Protector XL Hoods
Cut Style Left and Right Rear Cupsink Cutouts