missing translation for 'onlineSavingsMsg'
Learn More

Spilanthol (Technical Grade), TRC

Product Code. 30008894
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008894 1 mg 1mg
30003732 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008894 Supplier Toronto Research Chemicals Supplier No. TRCS6818501MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25394-57-4
Molecular Formula C14 H23 N O
Molecular Weight (g/mol) 221.34
Synonym (2E,6Z,8E)-N-(2-Methylpropyl)-2,6,8-decatrienamide, (E,E,Z)-N-(2-Methylpropyl)-2,6,8-decatrienamide, (E,E,Z)- N-Isobutyl-2,6,8-decatrienamide, Affinin, 2E,6Z,8E-Decatrienoic Acid N-Isobutylamide, N-Isobutyldeca-trans-2,cis-6,trans-8-trienamide;
IUPAC Name (2E,6E,8Z)-N-(2-methylpropyl)deca-2,6,8-trienamide
SMILES C\C=C/C=C/CC\C=C\C(=O)NCC(C)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H23NO/c1-4-5-6-7-8-9-10-11-14(16)15-12-13(2)3/h4-7,10-11,13H,8-9,12H2,1-3H3,(H,15,16)/b5-4-,7-6+,11-10+
Formula Weight 221.178
Chemical Name or Material Spilanthol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.