missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spike S1 protein Mouse anti-SARS-CoV-2, Unconjugated, Monoclonal Antibody, AcroBiosystems
Mouse Monoclonal Antibody
£374.00 - £2730.00
Specifications
| Antigen | Spike S1 protein |
|---|---|
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Hybridoma |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17146691
|
ACROBiosystems
S1N-S59A1-100ug |
100 μg |
£374.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17156691
|
ACROBiosystems
S1N-S59A1-1mg |
1 mg |
£2730.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Spike S1 protein Monoclonal antibody specifically detects Spike S1 protein in SARS-CoV-2 samples.Specifications
| Spike S1 protein | |
| Unconjugated | |
| RUO | |
| IgG1 | |
| -20°C |
| Monoclonal | |
| Hybridoma | |
| SARS-CoV-2 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title