missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spike RBD Antibody (2847A), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB11198J
This item is not returnable.
View return policy
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Western Blot, Flow Cytometry
Specifications
| Spike RBD | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| CHO-derived sars-cov-2 Spike RBD protein, Arg319-Phe541, Accession # YP_009724390.1 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry | |
| 2847A | |
| Western Blot, Flow Cytometry | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| SARS-CoV-2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction