missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spike RBD Antibody (1049349), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB11055F
This item is not returnable.
View return policy
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| Spike RBD | |
| Monoclonal | |
| FITC | |
| PBS | |
| Human embryonic kidney cell HEK293-derived SARS-CoV-2 Spike RBD, Val16-Pro681, Accession # YP_009724390.1 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry | |
| 1049349 | |
| Flow Cytometry, Immunocytochemistry | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| SARS-CoV-2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction