missing translation for 'onlineSavingsMsg'
Learn More

Spiclomazine, TRC

Product Code. 30411784
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411784 50 mg -
30417932 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30411784 Supplier Toronto Research Chemicals Supplier No. TRCS68176050MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24527-27-3
Molecular Formula C22H24ClN3OS2
Molecular Weight (g/mol) 446.03
Synonym Clospirazine, 8-[3-(2-Chlorophenothiazin-10-yl)propyl]-1-thia-4,8-diazaspiro[4.5]decan-3-one
IUPAC Name 8-[3-(2-chlorophenothiazin-10-yl)propyl]-1-thia-4,8-diazaspiro[4.5]decan-3-one
SMILES Clc1ccc2Sc3ccccc3N(CCCN4CCC5(CC4)NC(=O)CS5)c2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H24ClN3OS2/c23-16-6-7-20-18(14-16)26(17-4-1-2-5-19(17)29-20)11-3-10-25-12-8-22(9-13-25)24-21(27)15-28-22/h1-2,4-7,14H,3,8-13,15H2,(H,24,27)
Formula Weight 445.1049
Chemical Name or Material Spiclomazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.