missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SPICE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30409
This item is not returnable.
View return policy
Description
SPICE1 Polyclonal antibody specifically detects SPICE1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SPICE1 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| CCDC52, FLJ26064, SPICE, spindle and centriole associated protein 1 | |
| between 475 and 525 | |
| 20 μg | |
| Cell Biology, Cell Cycle and Replication | |
| 152185 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction