missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPIC Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535987
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, Raji, NCI-H929. Predicted reactivity: Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Controls the development of red pulp macrophages required for red blood cells recycling and iron homeostasis. Transcription factor that binds to the PU-box, a purine-rich DNA sequence (5'-GAGGA[AT]-3') that can act as a lymphoid-specific enhancer. Regulates VCAM1 gene expression.
Specifications
| SPIC | |
| Polyclonal | |
| Unconjugated | |
| Spic | |
| AU019198; C76795; Prf; Pu.1-related factor; SPIC; SPI-C; Spi-C transcription factor; Spi-C transcription factor (Spi-1/PU.1 related); Transcription factor Spi-C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 121599 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q8N5J4 | |
| Spic | |
| Recombinant protein encompassing a sequence within the center region of human Spi-C. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction