missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPIC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549851
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Spi-C protein.
Controls the development of red pulp macrophages required for red blood cells recycling and iron homeostasis. Transcription factor that binds to the PU-box, a purine-rich DNA sequence (5'-GAGGA[AT]-3') that can act as a lymphoid-specific enhancer. Regulates VCAM1 gene expression.
Specifications
| SPIC | |
| Polyclonal | |
| Unconjugated | |
| Spic | |
| AU019198; C76795; Prf; Pu.1-related factor; SPIC; SPI-C; Spi-C transcription factor; Spi-C transcription factor (Spi-1/PU.1 related); Transcription factor Spi-C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 121599 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N5J4 | |
| Spic | |
| Synthesized peptide derived from internal of human SPIC. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction