missing translation for 'onlineSavingsMsg'
Learn More

Spi-B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£169.00 - £255.00

Specifications

Antigen Spi-B
Dilution Immunoprecipitation 4-8 uL/mg of lysate
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18600896
View Documents
Novus Biologicals
NBP2-98958-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18608935
View Documents
Novus Biologicals
NBP2-98958-50ul
50 μg
£169.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Spi-B Polyclonal antibody specifically detects Spi-B in Human samples. It is validated for Immunoprecipitation
Specifications

Specifications

Spi-B
Immunoprecipitation
Unconjugated
Rabbit
Human
SPI-B, Spi-B transcription factor (Spi-1/PU.1 related), transcription factor Spi-B
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Spi-B.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunoprecipitation 4-8 uL/mg of lysate
Polyclonal
Purified
RUO
0.2 μm filtered solution in PBS
6689
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.