missing translation for 'onlineSavingsMsg'
Learn More

Spi-B Antibody [DyLight 488], Novus Biologicals™

Product Code. 30732076 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732076 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732076 Supplier Novus Biologicals Supplier No. AF7204K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Spi-B Polyclonal antibody specifically detects Spi-B in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spi-B
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias SPI-B, Spi-B transcription factor (Spi-1/PU.1 related), transcription factor Spi-B
Host Species Sheep
Immunogen E. coli-derived recombinant mouse Spi-B, Tyr18-Glu167 (Tyr110Phe), Accession # O35906
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6689
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.