missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPHK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528099
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, Molt-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SPHK1, along with SPHK2, catalyzes the phosphorylation of sphingosine producing the signaling molecule sphingosine-1-phosphate (S1P). S1P signals through G-protein coupled receptors and controls diverse cellular processes. Inhibitors of SPHKs are viewed as potential therapeutics for the treatment of cancer. SPHK1 catalyzes the phosphorylation of sphingosine to sphingosine 1 phosphate (S1P), an important signaling molecule with intra- and extracellular functions. Inside the cell S1P acts as a signaling molecule like other sphingolipid metabolites like ceramide and sphingosine. S1P has been implicated in regulating cell differentiation, calcium mobilization from intracellular stores, and apoptosis. The cell surface receptors for S1P are the EDG family of G protein-coupled receptors (S1P Receptors).
Specifications
| SPHK1 | |
| Polyclonal | |
| Unconjugated | |
| SPHK1 | |
| 1110006G24Rik; Acetyltransferase SPHK1; RP23-193A16.4; SK 1; Sk1; SKase; Sphingosine kinase 1; SPHK; SPHK1; SPK; SPK 1; Spk1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8877 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.17 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9NYA1 | |
| SPHK1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SPHK1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction