missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPHK1 Monoclonal Antibody (647706)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561567

Product Code. 30536230

  • £357.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

SPHK1, along with SPHK2, catalyzes the phosphorylation of sphingosine producing the signaling molecule sphingosine-1-phosphate (S1P). S1P signals through G-protein coupled receptors and controls diverse cellular processes. Inhibitors of SPHKs are viewed as potential therapeutics for the treatment of cancer. SPHK1 catalyzes the phosphorylation of sphingosine to sphingosine 1 phosphate (S1P), an important signaling molecule with intra- and extracellular functions. Inside the cell S1P acts as a signaling molecule like other sphingolipid metabolites like ceramide and sphingosine. S1P has been implicated in regulating cell differentiation, calcium mobilization from intracellular stores, and apoptosis. The cell surface receptors for S1P are the EDG family of G protein-coupled receptors (S1P Receptors).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPHK1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9NYA1
SPHK1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Sphingosine Kinase 1/SPHK1, Asp2-Leu398.
100 μg
Primary
Human
Antibody
IgG1
ELISA
647706
Unconjugated
SPHK1
1110006G24Rik; Acetyltransferase SPHK1; RP23-193A16.4; SK 1; Sk1; SKase; Sphingosine kinase 1; SPHK; SPHK1; SPK; SPK 1; Spk1
Mouse
Protein A/G
RUO
8877
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.