missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Sphingosine Kinase 2/SPHK2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£162.00 - £532.00

Specifications

Antigen Sphingosine Kinase 2/SPHK2
Dilution Western Blot : 1-5 μg/mL, ELISA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30067123
View Documents
Novus Biologicals™
NBP3-30877-100ug
100 μg
£532.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30067222
View Documents
Novus Biologicals™
NBP3-30877-25ug
25 μg
£162.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sphingosine Kinase 2/SPHK2 Polyclonal antibody specifically detects Sphingosine Kinase 2/SPHK2 in Human samples. It is validated for ELISA,Western Blot
Specifications

Specifications

Sphingosine Kinase 2/SPHK2
ELISA, Western Blot
Unconjugated
Rabbit
Apoptosis
Phosphate buffered saline
56848
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot : 1-5 μg/mL, ELISA
Polyclonal
Purified
RUO
Human
EC 2.7.1.91, SK 2, sphingosine kinase 2, sphingosine kinase type 2 isoform, SPK 2
Synthetic peptide derived from the N-terminal extension of the Sphingosine Kinase 2/SPHK2 protein.
Ammonium sulfate precipitation
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.