missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Sphingosine Kinase 2/SPHK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Sphingosine Kinase 2/SPHK2 |
|---|---|
| Dilution | Western Blot : 1:50-1:200, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067145
|
Novus Biologicals™
NBP3-30857-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066771
|
Novus Biologicals™
NBP3-30857-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sphingosine Kinase 2/SPHK2 Polyclonal antibody specifically detects Sphingosine Kinase 2/SPHK2 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Sphingosine Kinase 2/SPHK2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| Phosphate buffered saline | |
| 56848 | |
| IgG | |
| Antigen Affinity-purified |
| Western Blot : 1:50-1:200, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.1.91, SK 2, sphingosine kinase 2, sphingosine kinase type 2 isoform, SPK 2 | |
| Rabbits were immunized with a synthetic peptide derived from the Sphingosine Kinase 2/SPHK2 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title