missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Sphingosine Kinase 1/SPHK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29344
This item is not returnable.
View return policy
Description
Sphingosine Kinase 1/SPHK1 Polyclonal antibody specifically detects Sphingosine Kinase 1/SPHK1 in Human samples. It is validated for Immunoprecipitation
Specifications
| Sphingosine Kinase 1/SPHK1 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1 | |
| between 1 and 50 | |
| 100 μg | |
| Lipid and Metabolism | |
| 8877 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction