missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sphingosine Kinase 1/SPHK1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB110-40449
This item is not returnable.
View return policy
Description
Sphingosine Kinase 1/SPHK1 Polyclonal specifically detects Sphingosine Kinase 1/SPHK1 in Human, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Sphingosine Kinase 1/SPHK1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.1% Sodium Azide | |
| SPHK1 | |
| Synthetic 15 amino acid peptide from N-terminus of human Sphingosine Kinase 1/SPHK1. | |
| 0.05 mg | |
| Lipid and Metabolism | |
| 8877 | |
| Human, Monkey | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml | |
| EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific to the human Sphingosine Kinase Type 1 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction