missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sphingosine-1-phosphate Antibody (sonepcizumab), Alexa Fluor™ 532, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28006AF532
This item is not returnable.
View return policy
Description
Sphingosine-1-phosphate Monoclonal antibody specifically detects Sphingosine-1-phosphate in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Sphingosine-1-phosphate | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CD363, CD363 antigen, CHEDG1, D1S3362, ECGF1, EDG1, EDG-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, S1P receptor 1, S1P receptor Edg-1, s1p1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1 | |
| Sphingosine-1-phosphate | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| sonepcizumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction