Learn More
Sphingosine-1-phosphate Antibody (Expression DD patent anti-SIP) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28683JF669
Product Code. 30171970
  • £328.00 / 0.10mL
Description
Sphingosine-1-phosphate Monoclonal antibody specifically detects Sphingosine-1-phosphate in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sphingosine-1-phosphate
Monoclonal
Janelia Fluor 669
CD363, CD363 antigen, CHEDG1, D1S3362, ECGF1, EDG1, EDG-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, S1P receptor 1, S1P receptor Edg-1, s1p1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1
Sphingosine-1-phosphate
0.1 mL
Primary
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
Expression DD patent anti-SIP
50mM Sodium Borate
Human
Protein A purified
RUO
Human
Purified