missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Sphingomyelin synthase 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £532.00
Specifications
| Antigen | Sphingomyelin synthase 1 |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066942
|
Novus Biologicals™
NBP3-30869-100ug |
100 μg |
£532.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066865
|
Novus Biologicals™
NBP3-30869-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sphingomyelin synthase 1 Polyclonal antibody specifically detects Sphingomyelin synthase 1 in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| Sphingomyelin synthase 1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| Phosphate buffered saline | |
| 259230 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 5-10 μg/mL, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| MGC17342, MOBProtein Mob, phosphatidylcholine:ceramide cholinephosphotransferase 1, SMS1EC 2.7.8.27, sphingomyelin synthase 1Medulla oblongata-derived protein, TMEM23hmob33, Transmembrane protein 23MOB1 | |
| Rabbits were immunized with a synthetic peptide derived from the mouse Sphingomyelin synthase 1 protein | |
| Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title