missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPG7 Monoclonal Antibody (OTI1C1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525909
This item is not returnable.
View return policy
Description
SPG7 Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a mitochondrial metalloprotease protein that is a member of the AAA family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. Mutations in this gene cause autosomal recessive spastic paraplegia 7. Two transcript variants encoding distinct isoforms have been identified.
Specifications
| SPG7 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3ULF4, Q7TT47, Q9UQ90 | |
| SPG7 | |
| Human recombinant protein fragment corresponding to amino acids 300-573 of SPG7 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| OTI1C1 | |
| Unconjugated | |
| SPG7 | |
| AI452278; AU015315; CAR; cell adhesion regulator; cell matrix adhesion regulator; CMAR; paraplegin; paraplegin, isoform 1; PGN; spastic paraplegia 7 (pure and complicated autosomal recessive); spastic paraplegia 7 homolog (human); spastic paraplegia 7 protein; SPG5C; SPG7; SPG7 matrix AAA peptidase subunit, paraplegin; SPG7, paraplegin matrix AAA peptidase subunit | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 234847, 353231, 489657, 6687 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction