missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPG21 Monoclonal Antibody (2B11)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521907
This item is not returnable.
View return policy
Description
Peptide Sequence: PHKIRDIPVT IMDVFDQSAL STEAKEEMYK LYPNARRAHL KTGGNFPYLC RSAEVNLYVQ IHLLQFHGTK YAAIDPSMVS AEELEVQKGS LGISQE.
The protein encoded by this gene was identified by a two-hybrid screen using CD4 as the bait. It binds to the hydrophobic C-terminal amino acids of CD4 which are involved in repression of T cell activation. The interaction with CD4 is mediated by the noncatalytic alpha/beta hydrolase fold domain of this protein. It is thus proposed that this gene product modulates the stimulatory activity of CD4. At least three different transcript variants encoding two different isoforms have been found for this gene.
Specifications
| SPG21 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9NZD8 | |
| SPG21 | |
| SPG21 (NP_057714, 211 a.a. ~ 306 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 2B11 | |
| Unconjugated | |
| SPG21 | |
| acid cluster protein 33; ACP33; BM-019; C78576; D9Wsu18e; GL010; Maspardin; MAST; spastic paraplegia 21 (autosomal recessive, Mast syndrome); Spastic paraplegia 21 autosomal recessive Mast syndrome protein; Spastic paraplegia 21 autosomal recessive Mast syndrome protein homolog; spastic paraplegia 21 homolog (human); Spastic paraplegia 21 protein; SPG21 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51324 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction