missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SPG21 Antibody (2B11), Novus Biologicals™

Product Code. 18347919 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347919 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347919 Supplier Novus Biologicals™ Supplier No. H00051324M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SPG21 Monoclonal antibody specifically detects SPG21 in Human samples. It is validated for ELISA,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPG21
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2B11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_057714
Gene Alias Acid cluster protein 33, ACP33Spastic paraplegia 21 protein, GL010, MASPARDIN, MASTBM-019, spastic paraplegia 21 (autosomal recessive, Mast syndrome), Spastic paraplegia 21 autosomal recessive Mast syndrome protein
Host Species Mouse
Immunogen SPG21 (NP_057714, 211 a.a. ∽ 306 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PHKIRDIPVTIMDVFDQSALSTEAKEEMYKLYPNARRAHLKTGGNFPYLCRSAEVNLYVQIHLLQFHGTKYAAIDPSMVSAEELEVQKGSLGISQE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neurodegeneration
Primary or Secondary Primary
Gene ID (Entrez) 51324
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.