missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPG11 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76990
This item is not returnable.
View return policy
Description
SPG11 Polyclonal specifically detects SPG11 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SPG11 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Colorectal carcinoma-associated protein, FLJ21439spatacsin, KIAA1840DKFZp762B1512, spastic paraplegia 11 (autosomal recessive), Spastic paraplegia 11 protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 80208 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| AAI53880 | |
| SPG11 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human SPG11. The immunogen is located within amino acids 2360 - 2410 of SPG11. Amino Acid Squence: FEEISKKYKQHQPTD | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction