missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Grinding Vial Set, Large Vial, PC Cylinder, Low-Chrome Steel End Plugs and Impactor
Shop All SPEX Certiprep ProductsDescription
- Grinds one large sample at a time in the Large Freezer/Mill™
- Sample grinding capacity 5 g – 50 g.
- Specifically designed to grind electronic components for RoHS/WEEE compliance testing.
- End plugs and impactor are made of low-chrome ASTM 06 steel to eliminate chromium contamination during grinding.
- Withstands liquid nitrogen temperatures.
- Closed grinding vial prevents sample cross-contamination, retains volatiles, and controls hazardous or critical samples.
- Transparent plastic center cylinder allows visual checking of the grinding progress.
- low-chrome steel end plugs and impactor are autoclavable and reusable.
- Polycarbonate center cylinders are disposable or may be cleaned with hot water and detergent.
- Compatible with 6804 Large Extractor/Vial Opener.
Specifications
Specifications
| Capacity (English) | 0.03 to 5.07 oz. |
| Capacity (Metric) | 50 mL |
| For Use With (Equipment) | 6870 Freezer/Mill |
| For Use With (Application) | For Grinding Electronic Components |
| Material | Low-chrome Steel |
| Capacity | 1 to 50 mL (0.03 to 5.07 oz.) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?