missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPESP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562301
This item is not returnable.
View return policy
Description
Immunogen sequence: KQTEAPRMLP VVTESSTSPY VTSYKSPVTT LDKSTGIGIS TESEDVPQLS GETAIEKPEE FGKHPESWNN DDILKKILDI NSQVQQALLS DTSNPA Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 41%.
The encoded protein is a human alloantigen involved in sperm-egg binding and fusion.
Specifications
| SPESP1 | |
| Polyclonal | |
| Unconjugated | |
| Spesp1 | |
| 4921508E09Rik; AI428149; equatorial segment protein; ESP; Glycosylated 38 kDa sperm protein C-7/8; RGD1563468; Sperm equatorial segment protein 1; SP-ESP; Spesp1; spesp1 {ECO:0000250; UniProtKB:Q6UW49}; UNQ732/PRO1418 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 246777 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UW49 | |
| Spesp1 | |
| Recombinant protein corresponding to Human SPESP1. Recombinant protein control fragment (Product #RP-98143). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction