missing translation for 'onlineSavingsMsg'
Learn More

Spermine, TRC

Product Code. 30134351
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134351 1 g 1g
30135944 5 g 5g
17991765 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134351 Supplier Toronto Research Chemicals Supplier No. TRCS6805101G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-44-3
Molecular Formula C10 H26 N4
Molecular Weight (g/mol) 202.34
Synonym 1,4-Butanediamine, N1,N4-bis(3-aminopropyl)-;1,4-Butanediamine, N,N'-bis(3-aminopropyl)- (8CI,9CI), Spermine (6CI), 1,5,10,14-Tetraazatetradecane, 4,9-Diazadodecane-1,12-diamine, Gerontine, Musculamine, N,N'-Bis(3-aminopropyl)-1,4-butanediamine, N,N'-Bis(3-aminopropyl)-1,4-tetramethylenediamine, N,N'-Bis(3-aminopropyl)butanediamine, NSC 268508, Neuridine, Spermin
IUPAC Name N,N'-bis(3-aminopropyl)butane-1,4-diamine
SMILES NCCCNCCCCNCCCN

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H26N4/c11-5-3-9-13-7-1-2-8-14-10-4-6-12/h13-14H,1-12H2
Formula Weight 202.2157
Percent Purity >95%
Chemical Name or Material Spermine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.