missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spermine synthase Antibody (OTI3C9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74318F
This item is not returnable.
View return policy
Description
Spermine synthase Monoclonal specifically detects Spermine synthase in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Spermine synthase | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| SMS | |
| Full length recombinant protein of human Spermine synthase (NP_004586) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3C9 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 2.5.1.22, MRSRSpS, Snyder-Robinson X-linked mental retardation syndrome, Spermidine aminopropyltransferase, spermine synthase, SPMSYSRS | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6611.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction