missing translation for 'onlineSavingsMsg'
Learn More

Spermine synthase Antibody, Novus Biologicals™

Chicken Polyclonal Antibody

£288.00

Specifications

Antigen Spermine synthase
Dilution Western Blot 1:100-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18503604
View Documents
Novus Biologicals
NB100-75148-0.025mg
0.025 mg
£288.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Spermine synthase Polyclonal specifically detects Spermine synthase in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Spermine synthase
Western Blot
Unconjugated
RUO
Human
P52788
6611
The immunogen for this antibody was a sequence from 206a.a. to 335a.a. from Spermine synthase(a 368a.a. long protein)
Affinity Purified
Spermine synthase
Western Blot 1:100-1:2000
Polyclonal
Chicken
Core ESC Like Genes, Stem Cell Markers
PBS with No Preservative
EC 2.5.1.22, MRSRSpS, Snyder-Robinson X-linked mental retardation syndrome, Spermidine aminopropyltransferase, spermine synthase, SPMSYSRS
SMS
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.