Learn More
Spermine, 97%, Thermo Scientific Chemicals
£28.90 - £108.00
Chemical Identifiers
CAS | 71-44-3 |
---|---|
Molecular Formula | C10H26N4 |
Molecular Weight (g/mol) | 202.35 |
MDL Number | MFCD00008215 |
InChI Key | PFNFFQXMRSDOHW-UHFFFAOYSA-N |
Synonym | spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl |
PubChem CID | 1103 |
ChEBI | CHEBI:15746 |
IUPAC Name | N,N'-bis(3-aminopropyl)butane-1,4-diamine |
SMILES | NCCCNCCCCNCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11385325
|
Thermo Scientific Alfa Aesar
L19562.03 |
1 g |
£28.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11395325
|
Thermo Scientific Alfa Aesar
L19562.06 |
5 g |
£108.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Binds to the polyamine modulatory site of NMDA Spermine is essential for both normal and neoplastic tissue growth. It is involved in the modulation of calcium-dependent immune processes. It plays an important role in cellular proliferation and differentiation as well as inhibits neuronal nitric oxide synthase (nNOS).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBinds to the polyamine modulatory site of NMDA Spermine is essential for both normal and neoplastic tissue growth. It is involved in the modulation of calcium-dependent immune processes. It plays an important role in cellular proliferation and differentiation as well as inhibits neuronal nitric oxide synthase (nNOS).
Solubility
Soluble in chloroform, methanol, water and lower alcohols. Insoluble in ether, benzene and petroleum ether.
Notes
Air sensitive and hygroscopic. Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
Chemical Identifiers
71-44-3 | |
202.35 | |
PFNFFQXMRSDOHW-UHFFFAOYSA-N | |
1103 | |
N,N'-bis(3-aminopropyl)butane-1,4-diamine |
C10H26N4 | |
MFCD00008215 | |
spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl | |
CHEBI:15746 | |
NCCCNCCCCNCCCN |
Specifications
71-44-3 | |
150°C (5 mmHg) | |
C10H26N4 | |
UN3259 | |
Air sensitive; Hygroscopic | |
spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl | |
PFNFFQXMRSDOHW-UHFFFAOYSA-N | |
N,N'-bis(3-aminopropyl)butane-1,4-diamine | |
1103 | |
202.35 | |
Spermine |
28°C to 30°C | |
>110°C (230°F) | |
MFCD00008215 | |
1751791 | |
14,8743 | |
Soluble in chloroform,methanol,water and lower alcohols. Insoluble in ether,benzene and petroleum ether. | |
NCCCNCCCCNCCCN | |
202.35 | |
CHEBI:15746 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 200-754-2
missing translation for 'rtecsNumber' : EJ7175000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only